Learn More
Sino Biological Anti-BATF3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102826T04100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human BATF3, and purified by antigen affinity chromatography.
Catalog No. 50-241-0781