Learn More
Sino Biological Recombinant Anti-BCAM Antibody (HRP), Rabbit Monoclonal
Supplier:  Sino Biological 10238R002H100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human BCAM (rh BCAM; Catalog#10238-H08H; NP_005572.2; Met1-Ala547), and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-241-2002