Learn More
Biomatik Corporation Human B-Cell Leukemia/Lymphoma 2 (Bcl2) CLIA Kit
Supplier:  Biomatik Corporation EKU08913
Encompass_Preferred
Human B-Cell Leukemia/Lymphoma 2 (Bcl2) CLIA Kit for detecting Bcl2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.41-300ng/ml. Sensitivity: 0.17ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P10415. Gene ID: 596. Target Synonyms: B-Cell CLL/Lymphoma 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7219