Learn More
Biomatik Corporation Human Bcl2 Associated Athanogene 3 (BAG3) ELISA Kit
Supplier:  Biomatik Corporation EKU02677
Encompass_Preferred
Human Bcl2 Associated Athanogene 3 (BAG3) ELISA Kit for detecting BAG3 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O95817. Gene ID: 9531. Target Synonyms: BIS; CAIR1; BAG Family Molecular Chaperone Regulator 3; Bcl-2-binding protein Bis; Docking protein CAIR-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9102