Learn More
Biomatik Corporation Human Bcl2 Associated Death Promoter (BAD) ELISA Kit
Supplier:  Biomatik Corporation EKU11994
Encompass_Preferred
Human Bcl2 Associated Death Promoter (BAD) ELISA Kit for detecting BAD in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 31pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q92934. Gene ID: 572. Target Synonyms: BBC2; BCL2L8; BCL2-Antagonist Of Cell Death; Bcl-2-binding component 6; Bcl-2-like protein 8; Bcl-xL/Bcl-2-associated death promoter; Bcl2 antagonist of cell death. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6574