Learn More
Sino Biological Human BCL2 ELISA Pair Set 15PP (Sino Biological)
Catalog No. 502373643
Encompass_Preferred
Each vial contains 252 ng of recombinant Human Bcl-2. Reconstitute with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 5000 pg/mL is recommended.
Catalog No. 50-237-3643 Supplier Sino Biological Supplier No. SEK1019515
Each vial contains 252 ng of recombinant Human Bcl-2. Reconstitute with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 5000 pg/mL is recommended.