Learn More
Sino Biological Anti-Bcl-2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 202274T36100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human Bcl-2 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-241-2502