Learn More
Sino Biological Anti-Bcl-W Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10059RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Bcl-W / BCL2L2 (rh Bcl-W / BCL2L2; Catalog#10059-H08E; Q92843-1; Met 1-Thr 172). Bcl-W / BCL2L2 specific IgG was purified by Human Bcl-W / BCL2L2 affinity chromatography.
Catalog No. 50-240-8190