Learn More
Sino Biological Anti-BCL6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12083RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human BCL6 (rh BCL6; Catalog#12083-H30E; P41182; Met1-Met150). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4463