Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human BCLAF1 (aa 349-482) Control Fragment Recombinant Protein

Catalog No. RP89286
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP89286 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP89286 Supplier Invitrogen™ Supplier No. RP89286
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (90%), Rat (90%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

BCLAF1 is an intracellular, death promoting transcriptional repressor of survival genes that interacts with several members of the BCL2 family of proteins. It shows homology to the basic leucine zipper (bzip) and MYB DNA binding domains.From recent research it can be concluded that in case of apoptotic cells this protein localizes to dot-like structures throughout the nucleus, and redistributes to a zone near the nuclear envelope. When over expressed this protein induces apoptosis and can be suppressed by co-expression of BCL2 proteins. Also known as BTF, it triggers a proapoptotic signal, potentially helpful to overcome the resistance to STI571.BCLAF1 is found to express mostly in skeletal muscles.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NYF8
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 9774
Name Human BCLAF1 (aa 349-482) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 2610102K23Rik; 2700025J07Rik; 2810454G14Rik; 5730534O06Rik; Aa2-041; AI450190; AW556225; BCL2 associated transcription factor 1; bcl-2-associated transcription factor 1; BCL2-associated transcription factor 1; Bcl-2-associated transcription factor 1-like protein; BCLAF1; BCLAF1 and THRAP3 family member 1; bK211L9.1; BP-1; BTF; Btf-s; KIAA0164; mKIAA0164
Common Name BCLAF1
Gene Symbol BCLAF1
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LLDRGNTRDKEASKEKGSEKGRAEGEWEDQEALDYFSDKESGKQKFNDSEGDDTEETEDYRQFRKSVLADQGKSFATASHRNTEEEGLKYKSKVSLKGNRESDGFREEKNYKLKETGYVVERPSTTKDKHKEED
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.