Learn More
ACROBiosystems APC-Labeled Human BCMA, His Tag (BCA-HA2H4) is produced via conjugation of APC to Human BCMA, His Tag with a new generation site-specific technology under Star Staining labeling platform. Human BCMA, His Tag is expressed from human 293 cell
Supplier:  ACROBiosystems BCAHA2H4200TESTS
Encompass_Preferred
Tumor necrosis factor receptor superfamily member 17 (TNFRSF17) is also known as B-cell maturation protein (BCMA), CD antigen CD269, which is a member of the TNF-receptor superfamily. TNFRSF17 contains one TNFR-Cys repeat. TNFRSF17 is expressed in mature B-cells, but not in T-cells or monocytes. TNFRSF17 is receptor for TNFSF13B/BLyS/BAFF and TNFSF13/APRIL. TNFRSF17 promotes B-cell survival and plays a role in the regulation of humoral immunity. TNFRSF17 can activate NF-kappa-B and JNK.
Catalog No. 50-225-3849