Learn More
ACROBiosystems Alexa Fluor 555-Labeled Human BCMA / TNFRSF17 Protein, His TagStar Staining
Catalog No. 502330919
Encompass_Preferred
Alexa Fluor 555-Labeled Human BCMA, His Tag (BCA-HA2H7) is produced via conjugation of AF555 to Human BCMA, His Tag with a new generation site-specific technology under Star Staining labeling platform. Human BCMA, His Tag is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 54 (Accession # Q02223-1).
Catalog No. 50-233-0919 Supplier ACROBiosystems Supplier No. BCAHA2H7200UG
Alexa Fluor 555-Labeled Human BCMA, His Tag (BCA-HA2H7) is produced via conjugation of AF555 to Human BCMA, His Tag with a new generation site-specific technology under Star Staining labeling platform. Human BCMA, His Tag is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 54 (Accession # Q02223-1).