Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human BCMA/TNFRSF17 ELISA Kit

Catalog No. EH41RBX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH41RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EH41RBX10 Supplier Invitrogen™ Supplier No. EH41RBX10
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

ELISA

Human BCMA/TNFRSF17 quantitates human BCMA/TNFRSF17 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human BCMA/TNFRSF17.

The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor is preferentially expressed in mature B lymphocytes, and may be important for B cell development and autoimmune response. This receptor has been shown to specifically bind to the tumor necrosis factor (ligand) superfamily, member 13b (TNFSF13B/TALL-1/BAFF), and to lead to NF-kappaB and MAPK8/JNK activation. This receptor also binds to various TRAF family members, and thus may transduce signals for cell survival and proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q02223
Assay Range 0.048-12 ng/mL
Assay Sensitivity 0.048 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias B cell maturation antigen; B-cell maturation factor; B-cell maturation protein; BCM; BCMA; CD269; TNF receptor superfamily member 17; Tnfrsf13; Tnfrsf13a; Tnfrsf17; Tumor necrosis factor receptor superfamily member 17; tumor necrosis factor receptor superfamily, member 17B cell maturation antigen; B-cell maturation factor; B-cell maturation protein; BCM; BCMA; CD269; TNF receptor superfamily member 17; Tnfrsf13; Tnfrsf13a; Tnfrsf17; Tumor necrosis factor receptor superfamily member 17; tumor necrosis factor receptor superfamily, member 17;
Gene ID (Entrez) 608
Gene Symbol Tnfrsf17
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target BCMA
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.