Learn More
ACROBiosystems ACROBiosystems Human BCMA / TNFRSF17 Protein, Llama IgG2b Fc Tag, low endotoxin
Catalog No. 502106446
Encompass_Preferred
Recombinant Protein;1MG;Human BCMA, Llama IgG2b Fc Tag, low endotoxin (BCA-H5259) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 54 (Accession # Q02223-1).;This protein carries a llama IgG2b Fc tag at the C-terminus. The protein has a calculated MW of 33.7 kDa. The protein migrates as 40-47 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BAFF, His Tag (Cat. No. BAF-H5248) at 5 μg/mL (100 μL/well) can bind Human BCMA, Llama IgG2b Fc Tag (Cat. No. BCA-H5259) with a linear range of 0.02-0.4 ng/mL (QC tested).;Immobilized Human BCMA, Llama IgG2b Fc Tag (Cat. No. BCA-H5259) at 2 μg/mL (100 μL/well) can bind Anti-Human BCMA Mab with a linear range of 0.4-3 ng/mL (Routinely tested).
Catalog No. 50-210-6446 Supplier ACROBiosystems Supplier No. BCAH52591MG
Recombinant Protein;1MG;Human BCMA, Llama IgG2b Fc Tag, low endotoxin (BCA-H5259) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Ala 54 (Accession # Q02223-1).;This protein carries a llama IgG2b Fc tag at the C-terminus. The protein has a calculated MW of 33.7 kDa. The protein migrates as 40-47 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BAFF, His Tag (Cat. No. BAF-H5248) at 5 μg/mL (100 μL/well) can bind Human BCMA, Llama IgG2b Fc Tag (Cat. No. BCA-H5259) with a linear range of 0.02-0.4 ng/mL (QC tested).;Immobilized Human BCMA, Llama IgG2b Fc Tag (Cat. No. BCA-H5259) at 2 μg/mL (100 μL/well) can bind Anti-Human BCMA Mab with a linear range of 0.4-3 ng/mL (Routinely tested).