Learn More
BIOHIPPO Human BDG Microsample
SDP
Catalog No. 5000315092
Encompass
Human BDG Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of BDG in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 6 4 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human BDG Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human BDG Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human BDG biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15092 Supplier BIOHIPPO Supplier No. BHE15210599
Human BDG Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of BDG in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 6 4 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human BDG Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human BDG Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human BDG biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color