Learn More
FUJIFILM BIOSCIENCES INC ACF Human BDNF, Recombinant Protein, Shenandoah Biotechnology, 10 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 10001AF10UG
Encompass_Preferred
Brain-derived neurotrophic factor (BDNF) is a nerve growth factor that binds two receptors, the low-affinity nerve growth factor receptor (LNGFR) and the tropomyosin receptor kinase B (TrkB), to support neuron growth and survival. BDNF expression in the hippocampus is essential for long-term memory storage and learning. Human, mouse, rat, and pig BDNF are cross-reactive.
Catalog No. 50-130-5749