Learn More
Raybiotech Inc HUMAN BDNF ELISA
Supplier:  Raybiotech Inc ELHBDNF1
Encompass_Preferred
RayBio Human BDNF ELISA Kit for cell culture supernatants, plasma, and serum samples. Pre-coated 96-well microplate kit with additional reagents. Detection Range: 80 pg/ml - 16 ng/ml
Catalog No. 50-104-3489