Learn More
FUJIFILM Biosciences Human BDNF, Recombinant Protein, Shenandoah Biotechnology, 10 ug vial
Catalog No. 50990601
Encompass_Preferred
Brain-derived neurotrophic factor (BDNF) is a nerve growth factor that binds two receptors, the low-affinity nerve growth factor receptor (LNGFR) and the tropomyosin receptor kinase B (TrkB), to support neuron growth and survival. BDNF expression in the hippocampus is essential for long-term memory storage and learning. Human, mouse, rat, and pig BDNF are cross-reactive.
Catalog No. 50-990-601 Supplier FUJIFILM Biosciences Supplier No. 1000110UG
Brain-derived neurotrophic factor (BDNF) is a nerve growth factor that binds two receptors, the low-affinity nerve growth factor receptor (LNGFR) and the tropomyosin receptor kinase B (TrkB), to support neuron growth and survival. BDNF expression in the hippocampus is essential for long-term memory storage and learning. Human, mouse, rat, and pig BDNF are cross-reactive.