Learn More
ACROBiosystems Human BDNF, Tag Free (BDF-H5219) is expressed from human 293 cells (HEK293). It contains AA His 129 - Arg 247 (Accession # P23560-1).
Supplier:  ACROBiosystems BDFH521920UG
Encompass_Preferred
Brain-derived neurotrophic factor (BDNF) along with nerve growth factor (NGF), neurotrophin-3 (NT-3), and neurotrophin-4/5 (NT-4/5) are members of the neurotrophin family of trophic factors. The neurotrophins play essential roles in the development, survival, and function of a wide range of neurons in both the peripheral and central nervous systems. The neurotrophins interact with two cell surface receptors, the low affinity P75 receptor and the Trk family of high affinity tyrosine kinase receptors. NGF preferentially binds TrkA, BDNF and NT4/5 bind TrkB, and NT-3 binds TrkC (and TrkA to a lesser extent).
Catalog No. 50-225-3323