Learn More
Sino Biological Anti-Beclin 1/BECN1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 105332T40100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Beclin 1/BECN1, and purified by antigen affinity chromatography.
Catalog No. 50-240-8897