Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human beta Amyloid (1-42) PTD Recombinant Protein

Catalog No. 03112
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03-112 1 mg
03-111 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03-112 Supplier Invitrogen™ Supplier No. 03112
Add to Cart
Edge
Add to Cart

Recombinant Protein

Human beta Amyloid (1-42) PTD Recombinant Protein for Bioactivity

Beta amyloid is a 40-43 amino acid peptide cleaved from amyloid precursor protein by a protease, gamma-secretase. Beta Amyloid (1-40) (Amyloid beta 40 or Abeta 40) together with beta Amyloid (1-42) (Amyloid beta 42 or Abeta 42) are two major C-terminal variants of the amyloid beta protein, constituting the majority of amyloid beta proteins. Both the Amyloid beta 40 and Amyloid beta 42 alloforms are major components of the extracellular plaques found in Alzheimer's disease brain tissue.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05067
For Use With (Application) Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 351
Molecular Weight (g/mol) 4.5 kDa
Name Human beta Amyloid (1-42) PTD
Purification Method Purified
Quantity 1 mg
Storage Requirements -20°C
Regulatory Status RUO
Gene Alias A beta 1 42; A β 1 42; A4; AAA; AB42; Abeta; Abeta40; Abeta42; ABPP; AD1; Adap; AG; AICD-50; AICD-57; AICD-59; AID(50); AID(57); AID(59); Alpha-CTF; Alpha-secretase C-terminal fragment; Alzheimer disease amyloid A4 protein homolog; alzheimer disease amyloid protein; Amyloid; amyloid A4; Amyloid b; Amyloid beta; amyloid beta (A4) precursor protein; Amyloid beta 1 42; amyloid beta A4 protein; amyloid beta precursor protein; Amyloid intracellular domain 50; Amyloid intracellular domain 57; Amyloid intracellular domain 59; Amyloid precursor protein; Amyloid β; Amyloid-beta A4 protein; Amyloid-beta precursor protein; Amyloid-beta protein 40; Amyloid-beta protein 42; Amyloidogenic glycoprotein; APP; APP-C57; APP-C59; APP-C99; APPI; appican; Aβ 1 42; B APP42; BAPP42; beta-amyloid peptide; beta-amyloid peptide(1-40); beta-amyloid peptide(1-42); beta-amyloid precursor protein; betaApp; Beta-APP40; BetaAPP42; Beta-APP42; Beta-CTF; Beta-secretase C-terminal fragment; C31; C80; C83; C99; Cerebral vascular amyloid peptide; CTFgamma; Cvap; E030013M08Rik; Gamma-CTF(50); Gamma-CTF(57); Gamma-CTF(59); Gamma-secretase C-terminal fragment 50; Gamma-secretase C-terminal fragment 57; Gamma-secretase C-terminal fragment 59; N-APP; P3(40); P3(42); peptidase nexin-II; PN2; PN-II; PreA4; protease nexin II; protease nexin-II; S-APP-alpha; S-APP-beta; Soluble APP-alpha; Soluble APP-beta; testicular tissue protein Li 2; β APP42; βAPP42
Common Name Amyloid beta 42
Gene Symbol APP
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Sequence H2N-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-His-His-Gln-Lys-Leu-Val-Phe-PheAla-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-Ile-Ala-OH
Content And Storage -20°C
Shipping Condition Dry Ice
Expression System proprietary
Form Lyophilized
Protein Subtype Amyloid-Related Proteins, Neuropeptides
Research Category Neurobiology
Purity or Quality Grade ≥95% by HPLC
Show More Show Less
How do you recommend that beta amyloid peptide be reconstituted?

Recommendations for Beta Amyloid Peptide Reconstitution:
(1) To induce peptide aggregation for neurotoxicity studies: The appearance of toxicity has recently been shown to correlate to the extent of beta sheet structure (S. Wang et al. [2001] J. Biol. Chem. 276(45):42027-42034). Recommended preincubation is: Dissolve the lyophilized peptide in 0.1% (v/v) trifluoroacetic acid in water at 10 mg/mL. Dilute the peptide to 0.5-1.0 mg/mL with PBS (without Ca2+). Incubate at 25°C for 24-48 hrs (24-36 hrs is usually sufficient). Neurotoxic activity is usually observed at 30-100 micrograms/mL.
(2) To prepare peptide for studies which require minimal peptide aggregation: Dissolve the peptide at a concentration of 1 mg/mL in 100% HFIP (1,1,1,3,3,3-hexafluoro-2-propanol [Sigma-Aldrich Cat. No. 325244, 99.8% ACS reagent grade]). Incubate at RT for 2 hours. During the incubation, vortex the peptide solution several times at moderate speed, allowing the HFIP to cover as much of the surface area as possible. Dry down the HFIP/peptide solution under a gentle stream of nitrogen gas. Continue drying for an additional 10 minutes. Cap vial immediately. Resuspend the peptide in 100% DMSO. Incubate the peptide plus DMSO for 12 minutes at RT with periodic vortexing at moderate speed. Add 10 microliters of this DMSO/peptide solution dropwise to 10 mL of BSAT-DPBS (see formulation below) while vortexing at moderate speed. Peptides prepared in this manner have been used as standards in ELISA for the detection of beta amyloid in biological samples. When using this peptide in ELISA, it is important that the assay buffer used with the peptide standards has the same composition as the samples under investigation.
Buffer Formulations: DPBS Solution (10X Stock) (Gibco#14200-075) Dulbecco's PBS (DPBS w/o Mg2+, Ca2+) BSAT-DPBS Solution: 1X DPBS, pH 7.4 5% BSA 0.03% Tween-20

Note: The inclusion of a protease inhibitor (i.e. AEBSF [Sigma Cat. No. A-8456]) is recommended when the BSAT-BPBS solution is to be used for the analysis beta amyloid-containing biological samples. To prepare a 40 mM stock solution of AEBSF, add 100 mg of AEBSF to 10 mL DPBS, pH 7.4, containing 5% BSA. The stock solution should be diluted to a concentration of 1 mM AEBSF in BSAT-DPBS just prior to use.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.