Learn More
Raybiotech Inc Human/Mouse/Rat Beta-defensin 1 EIA Kit.
Supplier:  Raybiotech Inc EIABD11
Encompass_Preferred
Optimized for serum and cell culture media. Competition-based ELISA on a 96-well strip plate.
Catalog No. 50-194-8800