Learn More
ACROBiosystems Human Beta-NGF Protein, Tag Free
Catalog No. 502331053
Encompass_Preferred
Human Beta-NGF Protein, Tag Free (BEF-H5214) is expressed from human 293 cells (HEK293). It contains AA Ser 122 - Ala 241 (Accession # P01138).
Catalog No. 50-233-1053 Supplier ACROBiosystems Supplier No. BEFH5214500UG
Human Beta-NGF Protein, Tag Free (BEF-H5214) is expressed from human 293 cells (HEK293). It contains AA Ser 122 - Ala 241 (Accession # P01138).