Learn More
Biomatik Corporation Human Beta-1,3-N-Acetylglucosaminyltransferase 3 (b3GNT3) ELISA Kit
Supplier:  Biomatik Corporation EKU08411
Encompass_Preferred
Human Beta-1,3-N-Acetylglucosaminyltransferase 3 (b3GNT3) ELISA Kit for detecting b3GNT3 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y2A9. Gene ID: 10331. Target Synonyms: B3GAL-T8; B3GN-T3; B3GNT-3; TMEM3; beta3Gn-T3; Putative Type II Membrane Protein; Transmembrane Protein 3; Core 1 extending beta-1,3-N-acetylglucosaminyltransferase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5846