Learn More
Biomatik Corporation Human Beta-1,4-Galactosyltransferase 1 (b4GALT1) ELISA Kit
Supplier:  Biomatik Corporation EKU02690
Encompass_Preferred
Human Beta-1,4-Galactosyltransferase 1 (b4GALT1) ELISA Kit for detecting b4GALT1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.64ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P15291. Gene ID: 2683. Target Synonyms: GTB; B4-GALT1; GGTB2; GT1; Beta4Gal-T1; Lactose Synthase A Protein; N-Acetyllactosamine Synthase; Nal synthase; beta-N-acetylglucosamine beta-1,4-galactosyltransferase 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4723