Learn More
Biomatik Corporation Human Beta-1,4-N-Acetyl Galactosaminyl Transferase 2 (b4GALNT2) ELISA Kit
Supplier:  Biomatik Corporation EKU02691
Encompass_Preferred
Human Beta-1,4-N-Acetyl Galactosaminyl Transferase 2 (b4GALNT2) ELISA Kit for detecting b4GALNT2 in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8NHY0. Gene ID: 124872. Target Synonyms: SD; B4GALGT2; B4GALT; Cad; GALGT2; Sda; Sd (a) beta-1,4-GalNAc transferase; UDP-GalNAc:Neu5Aca2-3Galb-R b1,4-N-acetylgalactosaminyltransferase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7313