Learn More
Biomatik Corporation Human Beta-2-Microglobulin (b2M) CLIA Kit
Supplier:  Biomatik Corporation EKU08861
Encompass_Preferred
Human Beta-2-Microglobulin (b2M) CLIA Kit for detecting b2M in serum, plasma, urine, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 68.6-50,000pg/ml. Sensitivity: 24.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P61769. Gene ID: 567. Target Synonyms: BMG; B2MG. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8610