Learn More
Biomatik Corporation Human Beta-Crosslaps (bCTx) ELISA Kit
Supplier:  Biomatik Corporation EKU02700
Encompass_Preferred
Human Beta-Crosslaps (bCTx) ELISA Kit for detecting bCTx in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 98.77-8,000pg/ml. Sensitivity: 35.37pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: b-CTx; bCTXI; bCTX-I; B-Cr; BCL; Type I Collagen C-Telopeptide-Related Fraction. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9099