Learn More
Biomatik Corporation Human Beta-Endorphin (bEP) CLIA Kit
Supplier:  Biomatik Corporation EKU08639
Encompass_Preferred
Human Beta-Endorphin (bEP) CLIA Kit for detecting bEP in serum, plasma, cerebrospinal fluid, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.81-2000pg/ml. Sensitivity: 2.89pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Accession Number: P01189. Gene ID: 5443. Target Synonyms: b-EP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5790