Learn More
Biomatik Corporation Human Betaine Homocysteine Methyltransferase (BHMT) ELISA Kit
Supplier:  Biomatik Corporation EKU02706
Encompass_Preferred
Human Betaine Homocysteine Methyltransferase (BHMT) ELISA Kit for detecting BHMT in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q93088. Gene ID: 635. Target Synonyms: Betaine—homocysteine S-methyltransferase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4724