Learn More
BIOHIPPO Human BGN Microsample
SDP
Catalog No. 5000314700
Encompass
Human BGN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of BGN in Serum plasma and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 128 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human BGN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human BGN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human BGN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14700 Supplier BIOHIPPO Supplier No. BHE15211715
Human BGN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of BGN in Serum plasma and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 128 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human BGN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human BGN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human BGN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color