Learn More
Sino Biological Anti-BID Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 10468MM10F100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human BID (rh BID; Catalog#10468-HNCE; P55957-1; Met 1-Asp 195) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-240-8476