Learn More
Sino Biological Anti-BID Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10468MM1020
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human BID (rh BID; Catalog#10468-HNCE; P55957-1; Met 1-Asp 195). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-9149