Learn More
Biomatik Corporation Human Big Dynorphin (Dyn) ELISA Kit
Supplier:  Biomatik Corporation EKU02718
Encompass_Preferred
Human Big Dynorphin (Dyn) ELISA Kit for detecting Dyn in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 6.17-500pg/ml. Sensitivity: 2.65pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01213. Gene ID: 5173. Target Synonyms: B-DYN; Biocytin 13 Dynorphin A 1-13 Amide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5829