Learn More
Abcam Human Biglycan Antibody Pair - BSA and Azide free, 10 x 96 Tests
Catalog No. 502663157
Encompass_Preferred
Human Biglycan Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human Biglycan.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-266-3157 Supplier Abcam Supplier No. AB25665010X96TESTS
Human Biglycan Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human Biglycan.
The product is subject to the following: Abcam Restricted Use Statement