Learn More
Raybiotech Inc Human Biglycan ELISA Kit
Supplier:  Raybiotech Inc ELHBGN1
Encompass_Preferred
For serum, plasma, and cell culture supernatant samples. Sensitivity: 0.51 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human Biglycan. Other species not determined.
Catalog No. 50-194-9020