Learn More
Biomatik Corporation Human Biliverdin Reductase A (BLVRA) ELISA Kit
Supplier:  Biomatik Corporation EKU02723
Encompass_Preferred
Human Biliverdin Reductase A (BLVRA) ELISA Kit for detecting BLVRA in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.274ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P53004. Gene ID: 644. Target Synonyms: BLVR; BVR; Biliverdin-IX alpha-reductase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9098