Learn More
Meso Scale Discovery R-PLEX Human BIM/Bcl2-L-11 Antibody Set, 5 plates
SDP
Catalog No. NC1836299
Encompass
The R-PLEX platform offers matched sets of antibodies and calibrators for a wide menu of biomarkers. The R-PLEX Human BIM/Bcl2-L-11 Antibody Set can be used with other MSD components to build assays that measure BIM/Bcl2-L-11. These assays can measure BIM/Bcl2-L-11 alone or in multiplex panels with other biomarkers.
Catalog No. NC1836299 Supplier Meso Scale Discovery Supplier No. F217A3
The R-PLEX platform offers matched sets of antibodies and calibrators for a wide menu of biomarkers. The R-PLEX Human BIM/Bcl2-L-11 Antibody Set can be used with other MSD components to build assays that measure BIM/Bcl2-L-11. These assays can measure BIM/Bcl2-L-11 alone or in multiplex panels with other biomarkers.