Learn More
Abcam Recombinant Human BIN1 protein, 100UG
Supplier:  Abcam AB98238100UG
Encompass_Preferred
Recombinant Human BIN1 protein is a Human Full Length protein, in the 1 to 439 aa range, expressed in Escherichia coli, with >90% purity and suitable for SDS-PAGE, MS.
Protein/ Peptide/ Enzyme Synonyms: AMPHL, BIN1, Myc box-dependent-interacting protein 1, Amphiphysin II, Amphiphysin-like protein, Box-dependent myc-interacting protein 1, Bridging integrator 1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2199979