Learn More
Sino Biological Anti-BLMH Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 201156T32100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human BLMH fragment, and purified by antigen affinity chromatography.
Catalog No. 50-240-9547