Learn More
Sino Biological Anti-BLMH Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11333RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human BLMH / Bleomycin Hydrolase (rh BLMH; Catalog#11333-H07E; NP_000377.1; Ser2-Glu455). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8447