Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human BLOC1S1 (aa 33-147) Control Fragment Recombinant Protein

Catalog No. RP91631
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP91631 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP91631 Supplier Invitrogen™ Supplier No. RP91631
  • $283.00
    $212.65 / Each of 1

    Save $70.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54395. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

BLOC1S1 is a component of the ubiquitously expressed BLOC1 multisubunit protein complex. BLOC1 is required for normal biogenesis of specialized organelles of the endosomal-lysosomal system, such as melanosomes and platelet dense granules (Starcevic and Dell'Angelica, 2004 ).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P78537
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 2647
Name Human BLOC1S1 (aa 33-147) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias AI839753; biogenesis of lysosomal organelles complex 1 subunit 1; biogenesis of lysosomal organelles complex-1, subunit 1; biogenesis of lysosome-related organelles complex 1 subunit 1; biogenesis of lysosome-related organelles complex 1 subunit 1-like; biogenesis of lysosome-related organelles complex-1, subunit 1; BLOC subunit 1; BLOC-1 subunit 1; BLOC1S1; BLOS1; BORCS1; GCN5 (general control of amino-acid synthesis, yeast, homolog)-like 1; GCN5 general control of amino acid synthesis-like 1; GCN5 general control of amino-acid synthesis 5-like 1; Gcn5l1; GCN5-like protein 1; general control of amino acid synthesis, yeast homolog-like 1; general control of amino acid synthesis-like 1; MICoA; MTA1-interacting coactivator; Protein RT14; RT14; si:dkey-102c8.4; Unknown (protein for MGC:137062)
Common Name BLOC1S1
Gene Symbol Bloc1s1
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LLKEHQAKQNERKELQEKRRREAITAATCLTEALVDHLNVGVAQAYMNQRKLDHEVKTLQVQAAQFAKQTGQWIGMVENFNQALKEIGDVENWARSIELDMRTIATALEYVYKGQ
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.