Learn More
Biomatik Corporation Human Block Of Proliferation 1 (BOP1) ELISA Kit
Supplier:  Biomatik Corporation EKU12074
Encompass_Preferred
Human Block Of Proliferation 1 (BOP1) ELISA Kit for detecting BOP1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q14137. Gene ID: 23246. Target Synonyms: Ribosome biogenesis protein BOP1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8040