Learn More
Biomatik Corporation Human B-Lymphocyte Activation Antigen B7-1 (LAB7-1) ELISA Kit
Supplier:  Biomatik Corporation EKU09061
Encompass_Preferred
Human B-Lymphocyte Activation Antigen B7-1 (LAB7-1) ELISA Kit for detecting LAB7-1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 33pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P33681. Gene ID: 941. Target Synonyms: CD80; CD28LG; CD28LG1; BB1; B7-1 Antigen; T-lymphocyte activation antigen CD80; CTLA-4 counter-receptor B7.1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6230