Learn More
Biomatik Corporation Human B-Lymphoid Tyrosine Kinase (BLK) ELISA Kit
Supplier:  Biomatik Corporation EKU02731
Encompass_Preferred
Human B-Lymphoid Tyrosine Kinase (BLK) ELISA Kit for detecting BLK in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.071ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P51451. Gene ID: 640. Target Synonyms: Tyrosine-protein kinase BLK; p55-Blk. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4948