Learn More
Raybiotech Inc Human BMP-15 (GDF-9B) ELISA Kit
Supplier:  Raybiotech Inc ELHBMP151
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.6 ng/ml . Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human BMP-15. It shows no cross-reactivity with human BMP-1, -2, -3, -4, -5, -6, -7, -8, or -10.
Catalog No. 50-194-9025