Learn More
Bon Opus Biosciences Recombinant Human BMP-2 (C-His) 50ug
SDP
Supplier:  Bon Opus Biosciences BP104350UG
Encompass_Preferred
Recombinant Human BMP-2 (C-His) is produced by E.coli. The target gene encoding Arg289-Arg396 is expressed with a C-His tag. Accession# NP_001191.1. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1043
Catalog No. 50-217-2642