Learn More
Raybiotech Inc Human BMP-2 ELISA Kit
Supplier:  Raybiotech Inc ELHBMP21
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 45 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: In this ELISA format, the antibody pair shows less than 1% cross-reactivity with rhBMP-3, rhBMP-4, rhBMP-5, rhBMP-6 or rhBMP-7.
Catalog No. 50-194-9026