Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human BMP-4, Animal-Free Recombinant Protein, PeproTech®

Catalog No. 502558504
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 μg
250 μg
50 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-255-8504 250 μg
AF12005ET2U 2 μg
50-162-8300 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-255-8504 Supplier Gibco™ Supplier No. AF12005ET250UG
Add to Cart
Add to Cart

Recombinant Protein

AF-120-05ET-1MG will be provided as 2 x 500 μg (AF-120-05ET-500UG). PeproTech's E.coli-derived BMP-4 is a fully active homodimeric protein consisting of two 107 amino acid subunits, which correspond to amino acids 303-408 of the full length BMP-4 precursor, and includes a N-terminal Met residue. The calculated molecular weight of Recombinant Human BMP-4 (E.coli-derived) is 24.2 kDa. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. The superfamily includes large families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. This particular family member plays an important role in the onset of endochondral bone formation in humans, and a reduction in expression has been associated with a variety of bone diseases, including the heritable disorder Fibrodysplasia Ossificans Progressiva. Alternative splicing in the 5' untranslated region of this gene has been described and three variants are described, all encoding an identical protein.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P12644
For Use With (Application) Control, Bioactivity
Format Lyophilized
Formulation Protein with no preservative
Gene ID (Entrez) 652
Molecular Weight (g/mol) 24.2 kDa
Name Human BMP-4, Animal-Free
Purification Method Purified
Quantity 250 μg
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/μg
Gene Alias BMP; BMP2B; BMP-2B; Bmp2b1; Bmp2b-1; Bmp4; Bmp-4; BOMPR4A; Bone morphogenetic protein; Bone morphogenetic protein 2B; bone morphogenetic protein 4; bone morphogenetic protein 4 preproprotein; DVR4; Dvr-4; MCOPS6; OFC11; ZYME
Common Name BMP4
Gene Symbol BMP4
Biological Activity Determined by its ability to induce alkaline phosphatase production by ATDC-5 cells. The expected ED50 for this effect is 5-10 ng/ml
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MKKNKNCRRH SLYVDFSDVG WNDWIVAPPG YQAFYCHGDC PFPLADHLNS TNHAIVQTLV NSVNSSIPKA CCVPTELSAI SMLYLDEYDK VVLKNYQEMV VEGCGCR
Content And Storage -20°C
Activity Determined by its ability to induce alkaline phosphatase production by ATDC-5 cells. The expected ED50 for this effect is 5-10 ng/ml.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.