Learn More
Bon Opus Biosciences Recombinant Human BMP-4 (C-hFc) 50ug
SDP
Supplier:  Bon Opus Biosciences BP104450UG
Encompass_Preferred
Recombinant Human BMP-4 (C-hFc) is produced by Human Cells. The target gene encoding Ser293-Arg408 is expressed with a C-hFc tag. Accession# P12644. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1044
Catalog No. 50-217-2643